Vinyltrichlorosilane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T0407-25G
T0407-500G
T0407-100G
TCIAT0407-25GEA
17.6
EUR
TCIAT0407-25G
TCIAT0407-500G
TCIAT0407-100G
Vinyltrichlorosilane ≥98.0% (by GC)
Vinyltrichlorosilane
Formule:
C₂H₃Cl₃Si Kookpunt: 92°C Smeltpunt: –95 °C Dichtheid: 1,2426 g/cm³ (20 °C) Vlampunt: 18°C |
MDL Nummer:
MFCD00000482 CAS nummer: 75-94-5 EINECS: 200-917-8 UN: 1305 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...