Ethoxytrimethylsilane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Trimethyl(ethoxy)silane
T1394-25ML T1394-250ML
TCIAT1394-25MLEA 31.9 EUR
TCIAT1394-25ML TCIAT1394-250ML
Ethoxytrimethylsilane ≥98.0% (by GC)
Ethoxytrimethylsilane
Formule: C₅H₁₄OSi
Kookpunt: 75°C
Smeltpunt: –83 °C
Dichtheid: 0,7573 g/cm³ (20 °C)
Vlampunt: –18°C
MDL Nummer: MFCD00009069
CAS nummer: 1825-62-3
EINECS: 217-370-6
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51728be01ee1bdf191b77338f2bb080ec2d0d749&vid=51728be01ee1bdf191b77338f2bb080ec2d0d749&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %