Indazole ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

I0278-1G I0278-5G
TCIAI0278-1GEA 27.5 EUR
TCIAI0278-1G TCIAI0278-5G
Indazole ≥99.0% (by GC)
Indazole
Formule: C₇H₆N₂
Kookpunt: 270 °C (1013 hPa)
Smeltpunt: 147…149 °C
Dichtheid: 1,32 g/cm³ (20 °C)
MDL Nummer: MFCD00005691
CAS nummer: 271-44-3
EINECS: 205-978-4
Merck Index: 14,04936
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c55c87bf69c708b411cba23ef768f62c372b86c&vid=5c55c87bf69c708b411cba23ef768f62c372b86c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 146.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR