Phenylpropiolic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid

P0610-25G P0610-5G
TCIAP0610-25GEA 207 EUR
TCIAP0610-25G TCIAP0610-5G
Phenylpropiolic acid ≥98.0% (by titrimetric analysis)
Phenylpropiolic acid
Formule: C₉H₆O₂
MW: 146.15 g/mol
Kookpunt: 294
MDL Nummer: MFCD00004361
CAS nummer: 637-44-5
EINECS: 211-285-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb2406578d2aef09fda849828371685b915418b4&vid=fb2406578d2aef09fda849828371685b915418b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 137.0 to 140.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR