Ethyl phenylpropiolate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P0814-25G P0814-5G
TCIAP0814-25GEA 134 EUR
TCIAP0814-25G TCIAP0814-5G
Ethyl phenylpropiolate ≥97.0% (by GC)
Ethyl phenylpropiolate
Formule: C₁₁H₁₀O₂
Kookpunt: 103°C
Smeltpunt: 60 °C
Dichtheid: 1,05 g/cm³ (20 °C)
Vlampunt: 110°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009185
CAS nummer: 2216-94-6
EINECS: 218-703-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f39d5287942192ddce1030631f1c597eb88225c3&vid=f39d5287942192ddce1030631f1c597eb88225c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR