Methyl phenylpropiolate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Methyl Phenylpropiolate, Phenylpropiolic acid methyl ester
M2080-25G
M2080-5G
TCIAM2080-25GEA
450
EUR
TCIAM2080-25G
TCIAM2080-5G
Methyl phenylpropiolate ≥98.0% (by GC)
Methyl phenylpropiolate
Formule:
C₁₀H₈O₂ MW: 160.17 g/mol Kookpunt: 103°C Smeltpunt: 26 Dichtheid: 1.083 |
MDL Nummer:
MFCD00041685 CAS nummer: 4891-38-7 |
|
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...