Propiolic acid ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
P0497-25G
P0497-5G
TCIAP0497-25GEA
302
EUR
TCIAP0497-25G
TCIAP0497-5G
Propiolic acid ≥97.0% (by GC, titration analysis)
Propiolic acid
Formule:
C₃H₂O₂ Kookpunt: 58°C Smeltpunt: 18 °C Dichtheid: 1,136 g/cm³ (20 °C) Vlampunt: 58°C |
MDL Nummer:
MFCD00004360 CAS nummer: 471-25-0 EINECS: 207-437-8 UN: 2920 ADR: 8,II Merck Index: 14,07821 |
|
Specification Test Results
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...