Ethyl crotonate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0418-25ML C0418-500ML
TCIAC0418-25MLEA 30.8 EUR
TCIAC0418-25ML TCIAC0418-500ML
Ethyl crotonate ≥98.0% (by GC)
Ethyl crotonate
Formule: C₆H₁₀O₂
Kookpunt: 143 °C (1013 hPa)
Smeltpunt: –79 °C
Dichtheid: 0,9208 g/cm³ (20 °C)
Vlampunt: 39°C
MDL Nummer: MFCD00009289
CAS nummer: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 14,02597
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87f42fde61a41a0e6d5de01456b3cb8fa56165a5&vid=87f42fde61a41a0e6d5de01456b3cb8fa56165a5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %