Tris(diethylamino)phosphine ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

T1309-25ML T1309-5ML
TCIAT1309-25MLEA 128 EUR
TCIAT1309-25ML TCIAT1309-5ML
Tris(diethylamino)phosphine ≥90.0% (by GC)
Tris(diethylamino)phosphine
Formule: C₁₂H₃₀N₃P
Kookpunt: 245°C
Dichtheid: 0,89 g/cm³ (20 °C)
Vlampunt: 26°C
MDL Nummer: MFCD00009041
CAS nummer: 2283-11-6
EINECS: 218-920-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97e24f43b66a4f76624136459122f3abfbc66d23&vid=97e24f43b66a4f76624136459122f3abfbc66d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR