Trimethyl orthoformate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
O0068-25ML O0068-500ML
TCIAO0068-25MLEA 18.7 EUR
TCIAO0068-25ML TCIAO0068-500ML
Trimethyl orthoformate ≥98.0% (by GC)
Trimethyl orthoformate
Formule: C₄H₁₀O₃
Kookpunt: 100°C
Smeltpunt: –53 °C
Dichtheid: 0,9691 g/cm³ (20 °C)
Vlampunt: 15°C
MDL Nummer: MFCD00008483
CAS nummer: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II
Merck Index: 14,06884
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77041bf6d4924558373f99b6c27efcdec399ff8b&vid=77041bf6d4924558373f99b6c27efcdec399ff8b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %