2-Benzylaminoethanol ≥95.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1130-25ML
B1130-500ML
TCIAB1130-25MLEA
36.3
EUR
TCIAB1130-25ML
TCIAB1130-500ML
2-Benzylaminoethanol ≥95.0% (by GC, titration analysis)
2-Benzylaminoethanol
Formule:
C₆H₅CH₂NHCH₂CH₂OH Kookpunt: 280°C Dichtheid: 1,1652 g/cm³ (20 °C) Vlampunt: 130°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002840 CAS nummer: 104-63-2 EINECS: 203-221-2 |
|
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...