(4-Chlorophenyl)acetyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2313-25G
C2313-100G
TCIAC2313-25GEA
80.3
EUR
TCIAC2313-25G
TCIAC2313-100G
(4-Chlorophenyl)acetyl chloride ≥98.0% (by GC, titration analysis)
(4-Chlorophenyl)acetyl chloride
Formule:
(C₆H₄Cl)CH₂COCl Kookpunt: 129°C Dichtheid: ~ 1 g/cm³ (20 °C) |
MDL Nummer:
MFCD00037111 CAS nummer: 25026-34-0 EINECS: 246-571-1 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...