(S)-(-)-Citronellal ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C1454-25ML C1454-5ML
TCIAC1454-25MLEA 125 EUR
TCIAC1454-25ML TCIAC1454-5ML
(S)-(-)-Citronellal ≥96.0% (by GC)
(S)-(-)-Citronellal
Formule: C₁₀H₁₈O
Kookpunt: 84 °C (1013 hPa)
Smeltpunt: –63 °C
Dichtheid: 0,85 g/cm³ (20 °C)
Vlampunt: 75°C
MDL Nummer: MFCD00075588
CAS nummer: 5949-05-3
EINECS: 227-707-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9153d67808308143c06750c8d64288138d60e4a&vid=c9153d67808308143c06750c8d64288138d60e4a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Specific rotation [a]20/D -13.0 to -18.0 deg (neat)