Toluene-3,4-dithiol ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0266-1G T0266-25G T0266-5G
TCIAT0266-1GEA 29.7 EUR
TCIAT0266-1G TCIAT0266-25G TCIAT0266-5G
Toluene-3,4-dithiol ≥97.0% (by GC, titration analysis)
Toluene-3,4-dithiol
Formule: C₇H₈S₂
Kookpunt: 137°C
Smeltpunt: 30…32 °C
Dichtheid: 1,18 g/cm³ (20 °C)
MDL Nummer: MFCD00004844
CAS nummer: 496-74-2
EINECS: 207-828-3
Merck Index: 14,09531
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da37b6f373026fec1bff13a5891221fa67feb206&vid=da37b6f373026fec1bff13a5891221fa67feb206&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %
Solubility in Sodium hydroxide solution almost transparency
Sensitiveness to pass test