4-Bromo-2,6-xylenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene
B2060-25G B2060-250G
TCIAB2060-25GEA 116 EUR
TCIAB2060-25G TCIAB2060-250G
4-Bromo-2,6-xylenol ≥98.0% (by GC)
4-Bromo-2,6-xylenol
Formule: C₈H₉BrO
MW: 201.06 g/mol
Kookpunt: 259
Smeltpunt: 79
MDL Nummer: MFCD00002314
CAS nummer: 2374-05-2
EINECS: 219-153-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4b100be84d10a83a145d8bbd4dd8800c1d49838&vid=b4b100be84d10a83a145d8bbd4dd8800c1d49838&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C