2-Bromo-4-fluorobenzyl alcohol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Bromo-4-fluorobenzyl Alcohol, (2-Bromo-4-fluorophenyl)methanol
B3417-5G
B3417-25G
TCIAB3417-5GEA
52.8
EUR
TCIAB3417-5G
TCIAB3417-25G
2-Bromo-4-fluorobenzyl alcohol ≥98.0% (by GC)
2-Bromo-4-fluorobenzyl alcohol
Formule:
C₇H₆BrFO MW: 205.03 |
MDL Nummer:
MFCD00672925 CAS nummer: 229027-89-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...