4-Butoxybenzyl alcohol ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B1042-25G
B1042-5G
TCIAB1042-25GEA
138
EUR
TCIAB1042-25G
TCIAB1042-5G
4-Butoxybenzyl alcohol ≥97.0% (by GC)
4-Butoxybenzyl alcohol
Formule:
C₁₁H₁₆O₂ Kookpunt: 169 - 171 °C (22 -rr) |
MDL Nummer:
MFCD00004657 CAS nummer: 6214-45-5 EINECS: 228-284-3 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Freezing point | 29.0 to 32.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...