(±)-α-Bromo-γ-butyrolactone ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1228-25G B1228-500G
TCIAB1228-25GEA 25.3 EUR
TCIAB1228-25G TCIAB1228-500G
(±)-α-Bromo-γ-butyrolactone ≥95.0% (by GC)
(±)-α-Bromo-γ-butyrolactone
Formule: C₄H₅BrO₂
Kookpunt: 140°C
Dichtheid: 1,994 g/cm³ (20 °C)
MDL Nummer: MFCD00005387
CAS nummer: 5061-21-2
EINECS: 225-764-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd45c79b295b8d72243e2f063cea52ad79779ad&vid=7cd45c79b295b8d72243e2f063cea52ad79779ad&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 95.0 %