2-Methoxynicotinic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid
M2041-25G
M2041-5G
TCIAM2041-25GEA
172
EUR
TCIAM2041-25G
TCIAM2041-5G
2-Methoxynicotinic acid ≥98.0% (by GC, titration analysis)
2-Methoxynicotinic acid
Formule:
C₇H₇NO₃ MW: 153.14 g/mol |
MDL Nummer:
MFCD00085110 CAS nummer: 16498-81-0 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...