2-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
M2089-1G
M2089-5G
TCIAM2089-1GEA
83.6
EUR
TCIAM2089-1G
TCIAM2089-5G
2-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
2-Methylnicotinic acid
Formule:
C₇H₇NO₂ Smeltpunt: 227°C |
MDL Nummer:
MFCD00013449 CAS nummer: 3222-56-8 |
|
Specification Test Results
Appearance | White to Green to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...