6-Aminonicotinic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

A1486-1G A1486-5G
TCIAA1486-1GEA 27.5 EUR
TCIAA1486-1G TCIAA1486-5G
6-Aminonicotinic acid ≥98.0% (by titrimetric analysis)
6-Aminonicotinic acid
Formule: C₆H₆N₂O₂
Smeltpunt: 312°C
MDL Nummer: MFCD00006326
CAS nummer: 3167-49-5
EINECS: 221-630-4
Merck Index: 14,00456
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b04fea42d1a339f0657964868c3b6b8e744130c&vid=7b04fea42d1a339f0657964868c3b6b8e744130c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR