(4-Hydroxyphenyl)acetonitrile ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H0657-25G H0657-5G
TCIAH0657-25GEA 71.5 EUR
TCIAH0657-25G TCIAH0657-5G
(4-Hydroxyphenyl)acetonitrile ≥99.0% (by GC)
(4-Hydroxyphenyl)acetonitrile
Formule: C₈H₇NO
Kookpunt: 150°C
Smeltpunt: 68…71 °C
MDL Nummer: MFCD00002383
CAS nummer: 14191-95-8
EINECS: 238-046-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54e701b94aabc224220db16baeb7874de48b9ef&vid=978a7ef4b1506eb52fdec6bd0d235ab6815afb88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 68.0 to 74.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR