4-Amino-3-methylbenzonitrile ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A2107-1G A2107-5G
TCIAA2107-1GEA 36.3 EUR
TCIAA2107-1G TCIAA2107-5G
4-Amino-3-methylbenzonitrile ≥97.0% (by GC)
4-Amino-3-methylbenzonitrile
Formule: C₈H₈N₂
Kookpunt: 292 °C (1013 hPa)
Smeltpunt: 95 °C
MDL Nummer: MFCD02093969
CAS nummer: 78881-21-7
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a89c9d2cd6c770e6b1aa2d28dcad5c98e6ab19&vid=63a89c9d2cd6c770e6b1aa2d28dcad5c98e6ab19&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 91.0 to 95.0 deg-C