1-Butylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-butylpyridinium bromide, 1-n-Butylpyridinium bromide
B1743-25G
B1743-5G
TCIAB1743-25GEA
93.5
EUR
TCIAB1743-25G
TCIAB1743-5G
1-Butylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Butylpyridinium bromide
Formule:
C₉H₁₄BrN |
MDL Nummer:
MFCD00671496 CAS nummer: 874-80-6 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...