2-Iodobenzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

I0659-1G I0659-25G
TCIAI0659-1GEA 22 EUR
TCIAI0659-1G TCIAI0659-25G
2-Iodobenzonitrile ≥98.0% (by GC)
2-Iodobenzonitrile
Formule: C₇H₄IN
Kookpunt: 147°C
MDL Nummer: MFCD00079761
CAS nummer: 4387-36-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f4c09c8f2fb444a00d25bb4a7dde42404ff614d&vid=0f4c09c8f2fb444a00d25bb4a7dde42404ff614d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR