Chloramphenicol ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
C2255-250G C2255-25G
TCIAC2255-250GEA 247 EUR
TCIAC2255-250G TCIAC2255-25G
Chloramphenicol ≥98.0% (by HPLC, titration analysis)
Chloramphenicol
Formule: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Kookpunt: 645
Smeltpunt: 150
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078159
CAS nummer: 56-75-7
EINECS: 200-287-4
Merck Index: 14,02077
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01c92d1d40765c760227ae3370d8d1882fb1eced&vid=01c92d1d40765c760227ae3370d8d1882fb1eced&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C
Specific rotation [a]20/D +17.0 to +20.0 deg (C=5, EtOH)