Phenoxyacetyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
P0113-25G
P0113-500G
TCIAP0113-25GEA
37.4
EUR
TCIAP0113-25G
TCIAP0113-500G
Phenoxyacetyl chloride ≥98.0% (by GC, titration analysis)
Phenoxyacetyl chloride
Formule:
C₈H₇ClO₂ Kookpunt: 226°C Smeltpunt: 28…30 °C Dichtheid: 1,23 g/cm³ (20 °C) Vlampunt: 108°C |
MDL Nummer:
MFCD00000726 CAS nummer: 701-99-5 EINECS: 211-862-4 UN: 3265 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...