Bromophenol blue sodium salt

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

S0043-1G S0043-5G
TCIAS0043-1GEA 33 EUR
TCIAS0043-1G TCIAS0043-5G
Bromophenol blue sodium salt
Bromophenol blue sodium salt
Formule: C₁₉H₉Br₄NaO₅S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013793
CAS nummer: 34725-61-6
EINECS: 252-170-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=631bb19c87f709c5c36309aa854f6125fce19663&vid=631bb19c87f709c5c36309aa854f6125fce19663&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Absorbance min. 0.480 (5 mg/L, pH 5.0, 590.0 to 594.0 nm)
Solubility in Water almost transparency
Drying loss max. 7.0 %
Ignition residue (Sulfate) 10.0 to 10.8 % (after drying)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR