Bromothymol blue sodium salt
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate
S0044-25G S0044-5G
TCIAS0044-25GEA 206 EUR
TCIAS0044-25G TCIAS0044-5G
Bromothymol blue sodium salt
Bromothymol blue sodium salt
Formule: C₂₇H₂₇Br₂NaO₅S
Smeltpunt: > 200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077263
CAS nummer: 34722-90-2
EINECS: 252-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f69fed2cbfe6ad641d2c9d79336f3b541c12b17&vid=0f69fed2cbfe6ad641d2c9d79336f3b541c12b17&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Orange to Brown powder to crystal
Drying loss max. 7.0 %
Ignition residue (Sulfate) 10.7 to 11.6 % (after drying)
Transition interval (pH) (yellow)6.0 ~ 7.6 (blue)