Eosin B
Leverancier: TCI EUROPE
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TCIAE0204-25GEA
28.6
EUR
TCIAE0204-25G
Eosin B
Eosin B
Formule:
C₂₀H₆Br₂N₂Na₂O₉ MW: 624,06 g/mol Smeltpunt: > 310 °C |
MDL Nummer:
MFCD00005041 CAS nummer: 548-24-3 EINECS: 208-943-1 Merck Index: 12,03638 |
Specification Test Results
Appearance | Amber to Brown powder to crystal |
Lambda max. | 514.0 to 518.0 nm (H2O) |
Absorbance (E1%1cm) | min. 900 (H2O, 514.0 to 518.0 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...