Ethylenetetracarbonitrile ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Tetracyanoethylene

T0077-25G T0077-5G
TCIAT0077-25GEA 200 EUR
TCIAT0077-25G TCIAT0077-5G
Ethylenetetracarbonitrile ≥98.0% (by total nitrogen basis)
Ethylenetetracarbonitrile
Formule: C₆N₄
MW: 128.09 g/mol
Kookpunt: 125°C
MDL Nummer: MFCD00001850
CAS nummer: 670-54-2
EINECS: 211-578-0
UN: 3439
ADR: 6.1 I
Merck Index: 14,09195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ee34f46cf4eaa1f5732bf7fff0df1035e11b1c6&vid=8ee34f46cf4eaa1f5732bf7fff0df1035e11b1c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Amber to Dark green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 198.0 to 202.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR