1,3-Di-tert-butylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D2431-1ML
D2431-5ML
TCIAD2431-1MLEA
105
EUR
TCIAD2431-1ML
TCIAD2431-5ML
1,3-Di-tert-butylbenzene ≥98.0% (by GC)
1,3-Di-tert-butylbenzene
Formule:
C₁₄H₂₂ MW: 190.33 Vlampunt: 84°C |
MDL Nummer:
MFCD00008830 CAS nummer: 1014-60-4 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...