1-Ethylpyrrole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-Ethylpyrrole

E0444-1G E0444-5G
TCIAE0444-1GEA 44 EUR
TCIAE0444-1G TCIAE0444-5G
1-Ethylpyrrole ≥98.0% (by GC)
1-Ethylpyrrole
Formule: C₆H₉N
MW: 95.14 g/mol
Kookpunt: 131°C
MDL Nummer: MFCD00060633
CAS nummer: 617-92-5
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f0e57a61e86a6e0b507ca0e4b1aac207b4e181f&vid=4f0e57a61e86a6e0b507ca0e4b1aac207b4e181f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR