3,7-Dihydroxy-2-naphthoic acid ≥90.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D2516-25G D2516-5G
TCIAD2516-25GEA 229 EUR
TCIAD2516-25G TCIAD2516-5G
3,7-Dihydroxy-2-naphthoic acid ≥90.0% (by HPLC)
3,7-Dihydroxy-2-naphthoic acid
Formule: C₁₁H₈O₄
MDL Nummer: MFCD00010253
CAS nummer: 83511-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62f566793d9da8bdc4659cbe1deddc74e1134bd7&vid=62f566793d9da8bdc4659cbe1deddc74e1134bd7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 90.0 area%