Uric acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1H-Purine-2,6,8(3H,7H,9H)-trione

U0018-25G U0018-500G
TCIAU0018-25GEA 40.7 EUR
TCIAU0018-25G TCIAU0018-500G
Uric acid ≥98.0% (by titrimetric analysis)
Uric acid
Formule: C₅H₄N₄O₃
Smeltpunt: 436 °C
Dichtheid: 1,89 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005712
CAS nummer: 69-93-2
EINECS: 200-720-7
Merck Index: 14,09875
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e44e4ae619a5700c8d244f1a2feb88d84933d569&vid=e44e4ae619a5700c8d244f1a2feb88d84933d569&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR