1,2-Diethoxyethaan ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Diethyl glycol, Ethylene glycol diethyl ether
D0456-25ML D0456-500ML
TCIAD0456-25MLEA 17.6 EUR
TCIAD0456-25ML TCIAD0456-500ML
1,2-Diethoxyethaan ≥98.0% (by GC)
1,2-Diethoxyethaan
Formule: C₆H₁₄O₂
MW: 118.18 g/mol
Kookpunt: 121°C
Smeltpunt: -74
Vlampunt: 35°C
MDL Nummer: MFCD00009253
CAS nummer: 629-14-1
EINECS: 211-076-1
UN: 1153
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a20ee1e38ecf0fac886d757551ff9f94c67f920&vid=2a20ee1e38ecf0fac886d757551ff9f94c67f920&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %