3-Acetylbenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C1993-25G
C1993-5G
TCIAC1993-25GEA
94.6
EUR
TCIAC1993-25G
TCIAC1993-5G
3-Acetylbenzonitrile ≥98.0% (by GC)
3-Acetylbenzonitrile
Formule:
C₉H₇NO Kookpunt: 120 °C (5 -rr) Smeltpunt: 98…100 °C |
MDL Nummer:
MFCD00001806 CAS nummer: 6136-68-1 EINECS: 228-110-6 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...