2-(Butylamino)ethanol ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B1023-25ML
B1023-500ML
TCIAB1023-25MLEA
17.6
EUR
TCIAB1023-25ML
TCIAB1023-500ML
2-(Butylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(Butylamino)ethanol
Formule:
C₆H₁₅NO Kookpunt: 199°C Dichtheid: 0,89 g/cm³ (20 °C) Vlampunt: 77°C |
MDL Nummer:
MFCD00020570 CAS nummer: 111-75-1 EINECS: 203-904-5 UN: 2735 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...