(+)-Cinchonine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol
C0350-200G
C0350-25G
TCIAC0350-200GEA
301
EUR
TCIAC0350-200G
TCIAC0350-25G
(+)-Cinchonine ≥98.0% (by titrimetric analysis)
(+)-Cinchonine
Formule:
C₁₉H₂₂N₂O Smeltpunt: 261…264 °C Dichtheid: 1,2 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00064372 CAS nummer: 118-10-5 EINECS: 204-234-6 Merck Index: 14,02287 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 80.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +218.0 to +228.0 deg (C=0.5, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...