Bilirubine
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid
B0460-1G
B0460-100MG
TCIAB0460-1GEA
240
EUR
TCIAB0460-1G
TCIAB0460-100MG
Bilirubine
Bilirubine
Formule:
C₃₃H₃₆N₄O₆ Smeltpunt: > 360 °C Dichtheid: 1,31 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00005499 CAS nummer: 635-65-4 EINECS: 211-239-7 Merck Index: 14,01218 |
|
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Lambda max. | 450.0 to 454.0 nm (CHCl3) |
Molar extinction coefficient | 59,900 to 61,500 (CHCl3, 450.0 to 454.0 nm) |