(±)-sec-Butylamine ≥99.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (±)-2-Butylamine, (±)-2-Aminobutane
B0708-25ML B0708-500ML
TCIAB0708-25MLEA 17.6 EUR
TCIAB0708-25ML TCIAB0708-500ML
(±)-sec-Butylamine ≥99.0% (by GC, titration analysis)
(±)-sec-Butylamine
Formule: C₄H₁₁N
MW: 73.14 g/mol
Kookpunt: 63°C
Smeltpunt: -104
Vlampunt: –9°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008094
CAS nummer: 13952-84-6
EINECS: 237-732-7
UN: 2733
ADR: 3 II
Merck Index: 14,01544
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2378ca4baaefb9907bef4503fdabfddbf8936c&vid=32ecf1780a32feed0fbc8ae8d7f250b5d87486d4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %