(±)-2-Butaanol ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
B0705-25ML B0705-500ML
TCIAB0705-25MLEA 17.6 EUR
TCIAB0705-25ML TCIAB0705-500ML
(±)-2-Butaanol ≥99.0% (by GC)
(±)-2-Butaanol
Formule: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Kookpunt: 100°C
Smeltpunt: –115°C
Vlampunt: 24°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004569
CAS nummer: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3 III
Merck Index: 14,01541
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47347320002d3593ecb0b9319c821663510fdfdd&vid=b16f9f7c5badc0f8bd3cdf5b7175d907e0133f7c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Density (20deg.C) 0.8060 to 0.8080 g/mL