DL-Camphor ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0011-500G C0011-25G
TCIAC0011-500GEA 81.4 EUR
TCIAC0011-500G TCIAC0011-25G
DL-Camphor ≥95.0% (by GC)
DL-Camphor
Formule: C₁₀H₁₆O
Kookpunt: 204 °C (1013 hPa)
Smeltpunt: ∼180 °C
Dichtheid: 0,992 g/cm³ (25 °C)
Vlampunt: 66 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074738
CAS nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III
Merck Index: 14,01732
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e86f8c77b88c3e7702b13fe9ac2004003f549c5&vid=cdd8bbc59fd8a8484b4f0e9a56516c64b70bfef6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless powder to crystal
Purity (GC) min. 95.0 %
Melting point 172.0 to 180.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR