1,3-Dibromopropane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D0202-25G
D0202-500G
D0202-100G
TCIAD0202-25GEA
17.6
EUR
TCIAD0202-25G
TCIAD0202-500G
TCIAD0202-100G
1,3-Dibromopropane ≥98.0% (by GC)
1,3-Dibromopropane
Formule:
BrH₂CCH₂CH₂Br Kookpunt: 167°C Smeltpunt: –34 °C Dichtheid: 1,99 g/cm³ (20 °C) Vlampunt: 56°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000255 CAS nummer: 109-64-8 EINECS: 203-690-3 UN: 1993 ADR: 3,III Merck Index: 14,09713 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...