1-Bromopentane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0628-25ML B0628-500ML
TCIAB0628-25MLEA 18.7 EUR
TCIAB0628-25ML TCIAB0628-500ML
1-Bromopentane ≥98.0% (by GC)
1-Bromopentane
Formule: BrH₂C(CH₂)₃CH₃
Kookpunt: 129 °C (1013 hPa)
Smeltpunt: –95 °C
Dichtheid: 1,22 g/cm³ (20 °C)
Vlampunt: 31°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000267
CAS nummer: 110-53-2
EINECS: 203-776-0
UN: 1993
ADR: 3,III
Merck Index: 14,00602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b0307a6a068f0ae80f9101fc6bb9d68b6de2d3&vid=4f62845be97aef07bca2b70fa28b9debdf81a373&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR