Broomethaan ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B0588-25G
B0588-500G
TCIAB0588-25GEA
17.6
EUR
TCIAB0588-25G
TCIAB0588-500G
Broomethaan ≥99.0% (by GC)
Broomethaan
Formule:
BrH₂CCH₃ Kookpunt: 38°C Smeltpunt: –118 °C Dichtheid: 1,46 g/cm³ (20 °C) Vlampunt: -23°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00000232 CAS nummer: 74-96-4 EINECS: 200-825-8 UN: 1891 ADR: 6.1,II Merck Index: 14,03771 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...