Bromotrichloromethane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B0662-25G
B0662-500G
TCIAB0662-25GEA
17.6
EUR
TCIAB0662-25G
TCIAB0662-500G
Bromotrichloromethane ≥98.0% (by GC)
Bromotrichloromethane
Formule:
BrCl₃C Kookpunt: 104°C Smeltpunt: –6 °C Dichtheid: 2,01 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000783 CAS nummer: 75-62-7 EINECS: 200-886-0 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...