Caprinaldehyde ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Decylaldehyde, Decanal, Decyl aldehyde

D0032-25ML D0032-500ML
TCIAD0032-25MLEA 17.6 EUR
TCIAD0032-25ML TCIAD0032-500ML
Caprinaldehyde ≥97.0% (by GC)
Caprinaldehyde
Formule: C₁₀H₂₀O
Kookpunt: 208°C
Smeltpunt: -5°C
Dichtheid: 0,83 g/cm³ (20 °C)
Vlampunt: 94°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007031
CAS nummer: 112-31-2
EINECS: 203-957-4
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c72fe8d4c64164da5416bf899deccdc7c40f7c2e&vid=5091177eb02c783811e2a394a7a2e8632728124c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR