Pelargonaldehyde ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

N0296-25ML N0296-500ML N0296-100ML
TCIAN0296-25MLEA 17.6 EUR
TCIAN0296-25ML TCIAN0296-500ML TCIAN0296-100ML
Pelargonaldehyde ≥95.0% (by GC)
Pelargonaldehyde
Formule: C₉H₁₈O
Kookpunt: 192°C
Smeltpunt: –40 °C
Dichtheid: 0,83 g/cm³ (20 °C)
Vlampunt: 71°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007030
CAS nummer: 124-19-6
EINECS: 204-688-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9054ec2591a1549225287a72f4328c0c22d81071&vid=bed93227f80405d0b83bb1b3c4c83dde1b2c62e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR