Triethylmethylammonium chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M0852-25G
M0852-500G
TCIAM0852-25GEA
18.7
EUR
TCIAM0852-25G
TCIAM0852-500G
Triethylmethylammonium chloride ≥98.0% (by titrimetric analysis)
Triethylmethylammonium chloride
Formule:
C₇H₁₈ClN |
MDL Nummer:
MFCD00059972 CAS nummer: 10052-47-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...